missing translation for 'onlineSavingsMsg'
Learn More
Trilaurin, Thermo Scientific Chemicals Product Code.: 15485249

Trilaurin, Thermo Scientific Chemicals

Product Code. 15485249
5 g
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy

Product Code. 15485249

Brand: Thermo Scientific Alfa Aesar J62962.06

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Trilaurin is used in makeup products, creams and lotions, deodorants, suntan and sunscreen products, hair conditioners, and skin care and skin cleansing products. These are used in cosmetic products as thickening agents and emollients. It is used as a viscosity controlling agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Trilaurin is used in makeup products, creams and lotions, deodorants, suntan and sunscreen products, hair conditioners, and skin care and skin cleansing products. These are used in cosmetic products as thickening agents and emollients. It is used as a viscosity controlling agent.

Solubility
Soluble in water, ethylacetate- almost transparency.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 538-24-9
Molecular Formula C39H74O6
Molecular Weight (g/mol) 639.015
MDL Number MFCD00026559
InChI Key VMPHSYLJUKZBJJ-UHFFFAOYSA-N
Synonym trilaurin, glycerol trilaurate, glyceryl tridodecanoate, glyceryl trilaurate, glycerin trilaurate, lauric acid triglyceride, tridodecanoin, lauric acid triglycerin ester, trilauroylglycerol, laurin, tri
PubChem CID 10851
ChEBI CHEBI:77389
IUPAC Name 2,3-di(dodecanoyloxy)propyl dodecanoate
SMILES CCCCCCCCCCCC(=O)OCC(COC(=O)CCCCCCCCCCC)OC(=O)CCCCCCCCCCC

Specifications

Melting Point 47°C
Color White
Linear Formula [CH3(CH2)10COOCH2]2CHOCO(CH2)10CH3
Quantity 5 g
Beilstein 1730452
Solubility Information Soluble in water,ethylacetate- almost transparency.
Formula Weight 639.01
Physical Form Powder
Chemical Name or Material Trilaurin

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Trilaurin, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.