missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRIM44 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11511-1-AP-150UL
This item is not returnable.
View return policy
Description
This is a rabbit polyclonal antibody raised against full length of human TRIM44.
May play a role in the process of differentiation and maturation of neuronal cells. May regulate the activity of TRIM17.Specifications
TRIM44 | |
Polyclonal | |
Unconjugated | |
TRIM44 | |
DIPB, HSA249128, MC7, Protein DIPB, TRIM44, tripartite motif containing 44 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
362172, 54765, 80985 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.27 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q6QA27, Q96DX7, Q9QXA7 | |
Trim44 | |
TRIM44 Fusion Protein Ag2070 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |