missing translation for 'onlineSavingsMsg'
Learn More

Trimethoprim, 99.8%, MP Biomedicals™

Product Code. 11485482 Shop All MP Biomedicals Products
1 g
Click to view available options
Quantity:
1 g
5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 738-70-5
Molecular Formula C14H18N4O3
Molecular Weight (g/mol) 290.323
InChI Key IEDVJHCEMCRBQM-UHFFFAOYSA-N
Synonym 2,4-Diamino-5-[3,4,5-trimethoxy-benzyl]pyrimidine
PubChem CID 5578
ChEBI CHEBI:45924
IUPAC Name 5-[(3,4,5-trimethoxyphenyl)methyl]pyrimidine-2,4-diamine
SMILES COC1=CC(=CC(=C1OC)OC)CC2=CN=C(N=C2N)N

Specifications

Type Trimethoprim
Physical Form Powder
Color White
Quantity 1 g
Assay Percent Range 0.998
Formula Weight 290.3
Chemical Name or Material Trimethoprim
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Trimethoprim, 99.8%, MP Biomedicals™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.