missing translation for 'onlineSavingsMsg'
Learn More

Trimethoxyboroxine, Thermo Scientific Chemicals

Product Code. 15442997
Click to view available options
Quantity:
250 g
This item is not returnable. View return policy
This item is not returnable. View return policy

It can be used in agrochemical, pharmaceutical and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 102-24-9
Molecular Formula C3H9B3O6
Molecular Weight (g/mol) 173.53
MDL Number MFCD00005946
InChI Key ZFMOJHVRFMOIGF-UHFFFAOYSA-N
Synonym trimethoxyboroxine, trimethoxyboroxin, boroxin, trimethoxy, trimethoxycyclotriboroxane, 2,4,6-trimethoxyboroxin, triboron trimethyl hexaoxide, boroxin, 2,4,6-trimethoxy, boric acid h3b3o6 , trimethyl ester, trimethoxy-boroxi, trimethoxyboroxole
PubChem CID 66880
SMILES COB1OB(OC)OB(OC)O1

Specifications

Melting Point 10°C to 11°C
Density 1.216 g/mL
Boiling Point 130°C
Flash Point 14°C (57°F)
Odor Strong
Refractive Index 1.3996
Quantity 250 g
UN Number UN2924
Beilstein 1709158
Sensitivity Air and moisture sensitive
Solubility Information Reacts with water.
IUPAC Name 2,4,6-trimethoxy-1,3,5,2,4,6-trioxatriborinane
Formula Weight 173.53
Physical Form Liquid
Chemical Name or Material Trimethoxyboroxine
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'