missing translation for 'onlineSavingsMsg'
Learn More
Trimethyl 1,3,5-benzenetricarboxylate, 99%, Thermo Scientific Chemicals Product Code.: 10764841

Trimethyl 1,3,5-benzenetricarboxylate, 99%, Thermo Scientific Chemicals

Product Code. 10764841
25 g, Glass bottle
Click to view available options
Quantity:
25 g
100 g
250 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10764841

Brand: Thermo Scientific Acros 152070250

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2672-58-4
Molecular Formula C12H12O6
Molecular Weight (g/mol) 252.22
MDL Number MFCD00008434
InChI Key RGCHNYAILFZUPL-UHFFFAOYSA-N
Synonym trimethyl 1,3,5-benzenetricarboxylate, trimethyl trimesate, trimesic acid trimethyl ester, 1,3,5-benzenetricarboxylic acid, trimethyl ester, 1,3,5-benzenetricarboxylic acid trimethyl ester, trimethylbenzene-1,3,5-tricarboxylate, 1,3,5-trimethyl benzene-1,3,5-tricarboxylate, methyl 3,5-bis methoxycarbonyl benzoate, benzene-1,3,5-tricarboxylic acid trimethyl ester, timtec-bb sbb007766Show MoreShow Less
PubChem CID 75881
IUPAC Name trimethyl benzene-1,3,5-tricarboxylate
SMILES COC(=O)C1=CC(=CC(=C1)C(=O)OC)C(=O)OC

Specifications

Melting Point 144°C to 147°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula C6H3(CO2CH3)3
Quantity 25 g
Beilstein 09, 979
Formula Weight 252.22
Percent Purity 99%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material Trimethyl 1,3,5-benzenetricarboxylate
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0442721 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number152071000,152070250
Lot NumberA0395076 Certificate TypeCertificate of Analysis Date15/12/2022 Catalog Number152071000,152070250,152072500
Lot NumberA0398275 Certificate TypeCertificate of Analysis Date15/12/2022 Catalog Number152071000,152070250,152072500
Lot NumberA0407520 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number152071000,152070250,152072500
Lot NumberA0442721 Certificate TypeCertificate of Analysis Date10/06/2022 Catalog Number152071000,152070250,152072500
Product Identifier
  • Trimethyl 1,3,5-benzenetricarboxylate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Trimethyl 1,3,5-benzenetricarboxylate, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.