missing translation for 'onlineSavingsMsg'
Learn More

Trimethyl orthobenzoate, 97%, Thermo Scientific Chemicals

Product Code. 10184140
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Alkylation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 707-07-3
Molecular Formula C10H14O3
Molecular Weight (g/mol) 182.22
MDL Number MFCD00008474
InChI Key IECKAVQTURBPON-UHFFFAOYSA-N
Synonym trimethyl orthobenzoate, trimethoxymethyl benzene, methyl orthobenzoate, benzene, trimethoxymethyl, trimethylorthobenzoate, orthobenzoic acid, trimethyl ester, orthobenzoic acid trimethyl ester, alpha,alpha,alpha-trimethoxytoluene, trimethoxyphenylmethane, trimethyl orthobeznoate
PubChem CID 69720
IUPAC Name trimethoxymethylbenzene
SMILES COC(C1=CC=CC=C1)(OC)OC

Specifications

Color Colorless
Density 1.0600g/mL
Boiling Point 87°C to 88°C (7.0 mmHg)
Flash Point 82°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula C6H5C(OCH3)3
Refractive Index 1.4880 to 1.49
Quantity 10 g
Fieser 13,327; 04,548; 13,195; 01,1256; 02,454; 04,565; 06,657; 07,414; 08,524; 09,512; 10,455; 13,301
Specific Gravity 1.06
Formula Weight 182.22
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Trimethyl orthobenzoate, 97%
Show More Show Less
compliance-icons
Product Identifier
  • Trimethyl orthobenzoate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : Trimethyl orthobenzoate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'