missing translation for 'onlineSavingsMsg'
Learn More

Trimethylamine, 45% w/w aq. soln., Thermo Scientific Chemicals

Product Code. 11319408
missing translation for 'orderingAttributeHoverText'
Quantity:
500 mL
2.5 L
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

Trimethylamine solution used in research. As flavoring agents and adjuvants. Trimethylamine is used in the synthesis of choline, tetramethylammonium hydroxide, plant growth regulators, strongly basic anion exchange resins, dye leveling agents and a number of basic dyes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identifiants chimiques

CAS 75-50-3
Molecular Formula C3H9N
Molecular Weight (g/mol) 59.11
MDL Number MFCD00008327
InChI Key GETQZCLCWQTVFV-UHFFFAOYSA-N
Synonym trimethylamine, methanamine, n,n-dimethyl, dimethylmethaneamine, n-trimethylamine, trimethylamine solution, ch3 3n, trimethyl amine, trimethylamin, trimethyl-amine, fema number 3241
PubChem CID 1146
ChEBI CHEBI:18139
SMILES CN(C)C

Spécification

Melting Point -2°C
Color Colorless
Boiling Point 30°C
Flash Point −20°C (−4°F)
Odor Unpleasant
Quantity 2.5 L
UN Number UN1297
Beilstein 956566
Merck Index 14,9710
Solubility Information Soluble In Water,8.9e+005mg/L.
IUPAC Name N,N-dimethylmethanamine
Formula Weight 59.11
Concentration or Composition (by Analyte or Components) 45% w/w aq. soln.
Physical Form Liquid
Chemical Name or Material Trimethylamine
Afficher plus Afficher moins

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'