missing translation for 'onlineSavingsMsg'
Learn More

Trimethylamine, pure, 4.2M (33 wt.%) solution in ethanol, Thermo Scientific Chemicals

Specifications

Chemical Name or Material Trimethylamine
CAS 75-50-3, 64-17-5
Color Colorless
Density 0.7500g/mL
Boiling Point 50.0°C
View More Specs

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Trimethylamine
Colorless
50.0°C
C3H9N
1.3630 to 1.3660
04,43
0.75
Solubility in water: soluble. Other solubilities: soluble in alcohol,ether,benzene,toluene,,xylene,ethylbenzene and chloroform
CN(C)C
1146
59.11
Pure
75-50-3, 64-17-5
0.7500g/mL
−20°C
(CH3)3N
MFCD00008327
15,988
trimethylamine, methanamine, n,n-dimethyl, dimethylmethaneamine, n-trimethylamine, trimethylamine solution, ch3 3n, trimethyl amine, trimethylamin, trimethyl-amine, fema number 3241
GETQZCLCWQTVFV-UHFFFAOYSA-N
59.11
CHEBI:18139
32 to 34%
Liquid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.
Harmful if inhaled.
Harmful if swallowed.
May cause respiratory irritation.
Highly flammable liquid and vapor.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre

GHS Signal Word: Danger

missing translation for 'einecsNumber' : 200-875-0

missing translation for 'rtecsNumber' : PA0350000

missing translation for 'tsca' : TSCA

Product Identifier
  • Trimethylamine, 33 wt.% solution in ethanol

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3

Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H332-Harmful if inhaled.
  • H335-May cause respiratory irritation.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

Supplemental information
  • MIXTURE LIST-Contains : Trimethylamine, ethanol

SDS
Documents

Documents

Product Certifications

For Manufacturing and Laboratory Use. Not for beverage use