missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trimethylamine, pure, 7.3M (50 wt.%) aqueous solution
Tienda Thermo Scientific Chemicals Productos
Click to view available options
Quantity:
500 mL
1 L
2.5 L
10 L
Packaging:
Glass bottle
Plastic drum
Tamaño de la unidad:
1 L
10 L
2.5 L
500mL
Descripción
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Especificaciones
Especificaciones
| Chemical Name or Material | Trimethylamine |
| Color | Colorless |
| CAS | 75-50-3, 7732-18-5 |
| Boiling Point | 30.0°C to 100.0°C |
| Melting Point | -2.0°C |
| Molecular Formula | C3H9N |
| Linear Formula | (CH3)3N |
| MDL Number | MFCD00008327 |
| Density | 0.8600g/mL |
| Synonym | trimethylamine, methanamine, n,n-dimethyl, dimethylmethaneamine, n-trimethylamine, trimethylamine solution, ch3 3n, trimethyl amine, trimethylamin, trimethyl-amine, fema number 3241 |
| Mostrar más |
Seguridad y manipulación
Nombre del producto
- Trimethylamine, 45-50 wt.% aqueous solution
- Danger
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Flammable liquid Category 1
- Skin corrosion/irritation Category 1 B
- Specific target organ toxicity Category 3
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H335-May cause respiratory irritation.
- H302-Harmful if swallowed.
- H332-Harmful if inhaled.
- H224-Extremely flammable liquid and vapour.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
RUO – Research Use Only
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido