missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trimethylboroxine, 3.5M (50 wt.%) solution in THF, AcroSeal™, Thermo Scientific Chemicals
Click to view available options
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
CAS | 823-96-1, 109-99-9 |
Melting Point | -38.0°C |
Color | Colorless to Yellow |
Density | 0.8900g/mL |
Boiling Point | 78.0°C to 80.0°C |
Flash Point | −9°C |
Packaging | AcroSeal™ Glass Bottle |
Molecular Formula | C3H9B3O3 |
MDL Number | MFCD00013354 |
Quantity | 100 mL |
Show More |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
A0457188 | Certificate of Analysis | 31/01/2025 | 429191000 |
A0457188 | Certificate of Analysis | 04/06/2024 | 429191000 |
A0450016 | Certificate of Analysis | 13/11/2023 | 429191000 |
A0437916 | Certificate of Analysis | 04/10/2022 | 429191000 |
A0424200 | Certificate of Analysis | 19/11/2020 | 429191000 |
Safety and Handling




Product Identifier
- Trimethylboroxine, 50 wt% solution in THF
- Danger
- Carcinogenicity Category 2
- Serious eye damage/eye irritation Category 1
- Flammable liquid Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H225-Highly flammable liquid and vapour.
- H315-Causes skin irritation.
- H318-Causes serious eye damage.
- H335-May cause respiratory irritation.
- H336-May cause drowsiness or dizziness.
- H351-Suspected of causing cancer.
- EUH019-May form explosive peroxides.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- MIXTURE LIST-Contains : Tetrahydrofuran, Trimethylboroxine
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Trimethylboroxine, 3.5M (50 wt.%) solution in THF, AcroSeal™, Thermo Scientific Chemicals >
Spot an opportunity for improvement?Share a Content Correction