missing translation for 'onlineSavingsMsg'
Learn More
Trimethylboroxine, 50% w/w soln. in THF, Thermo Scientific Chemicals Product Code.: 15478008

Trimethylboroxine, 50% w/w soln. in THF, Thermo Scientific Chemicals

Product Code. 15478008
5 mL
Click to view available options
Quantity:
5 mL
25 mL
This item is not returnable. View return policy

Product Code. 15478008

Brand: Thermo Scientific Alfa Aesar H61464.AA

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Trimethylboroxine is used as a derivatizing agent for GLC analysis. It is used in a diverse array of areas, including as a polymerization additive. It is also used in the preparation of CBS catalysts for asymmetric reductions.

Solubility
Not miscible in water.

Notes
Moisture and light sensitive. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 823-96-1
Molecular Formula C3H9B3O3
Molecular Weight (g/mol) 125.53
MDL Number MFCD00013354
InChI Key GBBSAMQTQCPOBF-UHFFFAOYSA-N
Synonym trimethylboroxine, 2,4,6-trimethylboroxin, trimethylboroxin, boroxin, trimethyl, trimethyl-1,3,5,2,4,6-trioxatriborinane, trimethyboroxine, trimethyl boroxin, trimethylboroxaine, trimethyl boroxine, trimethyl-boroxine
PubChem CID 574072
IUPAC Name 2,4,6-trimethyl-1,3,5,2,4,6-trioxatriborinane
SMILES CB1OB(C)OB(C)O1

Specifications

Density 0.894
Quantity 5 mL
UN Number UN2924
Beilstein 1757008
Sensitivity Light and moisture sensitive
Solubility Information Not miscible in water.
Formula Weight 125.54
Concentration or Composition (by Analyte or Components) 50% w/w soln. in THF
Chemical Name or Material Trimethylboroxine

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberX25K050 Certificate TypeCertificate of Analysis Date11/11/2024 Catalog NumberH61464.AA,H61464.AC
Lot NumberX22K014 Certificate TypeCertificate of Analysis Date05/11/2024 Catalog NumberH61464.AA,H61464.AC
Lot NumberQ11I031 Certificate TypeCertificate of Analysis Date15/04/2022 Catalog NumberH61464.AA,H61464.AC
Lot Number10195397 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberH61464.AA,H61464.AC
Lot NumberS12A018 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberH61464.AA,H61464.AC
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Trimethylboroxine, 50% w/w solution in THF
Signal Word
  • Danger
Hazard Category
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
  • H336-May cause drowsiness or dizziness.
  • H351-Suspected of causing cancer.
  • EUH019-May form explosive peroxides.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : Trimethylboroxine, Tetrahydrofuran

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Trimethylboroxine, 50% w/w soln. in THF, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.