missing translation for 'onlineSavingsMsg'
Learn More

Trimethylolpropane triacrylate, stabilized, Thermo Scientific Chemicals

Product Code. 11337188
Click to view available options
Quantity:
100 g
500 g
2 kg

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15625-89-5
Molecular Formula C15H20O6
Molecular Weight (g/mol) 296.32
MDL Number MFCD00008628
InChI Key DAKWPKUUDNSNPN-UHFFFAOYSA-N
Synonym trimethylolpropane triacrylate, tmpta, viscoat 295, nk ester a tmpt, sartomer sr 351, ogumont t 200, saret 351, setalux uv 2241, ccris 92, unii-4b67kgl96s
PubChem CID 27423
ChEBI CHEBI:35028
IUPAC Name 2,2-bis(prop-2-enoyloxymethyl)butyl prop-2-enoate
SMILES CCC(COC(=O)C=C)(COC(=O)C=C)COC(=O)C=C

Specifications

Density 1.10 g/mL
Boiling Point >200°C (1 mmHg)
Flash Point 87°C (188°F)
Odor Pungent
Refractive Index 1.4735
Quantity 100 g
Solubility Information Miscible with water.
Formula Weight 296.31
Physical Form Liquid
Chemical Name or Material Trimethylolpropane triacrylate, Stabilized
compliance-icons compliance-icons
Product Identifier
  • Trimethylolpropane triacrylate
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
Hazard Statement
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
  • H319-Causes serious eye irritation.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : Trimethylolpropane triacrylate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.