missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trimethylolpropane triacrylate, stabilized, Thermo Scientific Chemicals
Click to view available options
Quantity:
100 g
500 g
2 kg
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Density | 1.10 g/mL |
Boiling Point | >200°C (1 mmHg) |
Flash Point | 87°C (188°F) |
Odor | Pungent |
Refractive Index | 1.4735 |
Quantity | 500 g |
Solubility Information | Miscible with water. |
Formula Weight | 296.31 |
Physical Form | Liquid |
Chemical Name or Material | Trimethylolpropane triacrylate, Stabilized |
Safety and Handling
Product Identifier
- Trimethylolpropane triacrylate
- Warning
- LONG-TERM AQUATIC HAZARD Chronic 1
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Skin sensitiser Category 1
- H315-Causes skin irritation.
- H317-May cause an allergic skin reaction.
- H319-Causes serious eye irritation.
- H410-Very toxic to aquatic life with long lasting effects.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- MIXTURE LIST-Contains : Trimethylolpropane triacrylate
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction