missing translation for 'onlineSavingsMsg'
Learn More

(Trimethylsilyl)diazomethane, technical, approx. 1.8 to 2.4M solution in hexanes, AcroSeal™, ACROS Organics™

Product Code. 10141095
Click to view available options
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle

 

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Alkylation reagent
TRUSTED_SUSTAINABILITY

Specifications

CAS 18107-18-1, 92112-69-1
CAS Min % 32.15, 62.77
CAS Max % 37.23, 67.85
Color Yellow
Density 0.6750g/mL
Boiling Point 96.0°C
Flash Point −23°C
Packaging AcroSeal™ Glass Bottle
Molecular Formula C4H10N2Si
Linear Formula (CH3)3SiCHN2
MDL Number MFCD00053946
Quantity 100 mL
Specific Gravity 0.675
Synonym trimethylsilyl diazomethane, trimethylsilyldiazomethane, diazomethyl trimethylsilane, tms-diazomethane, diazomethyl trimethyl silane, tmschn2, silane, diazomethyl trimethyl, unii-qi98hqo8c4, diazomethyl-trimethyl-silane, qi98hqo8c4
Solubility Information Solubility in water: insoluble
InChI Key ONDSBJMLAHVLMI-UHFFFAOYSA-N
SMILES C[Si](C)(C)C=[N+]=[N-]
IUPAC Name diazomethyl(trimethyl)silane
Molecular Weight (g/mol) 114.22
PubChem CID 167693
Formula Weight 114.22
Grade Technical
Physical Form Solution
Chemical Name or Material (Trimethylsilyl)diazomethane
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • (Trimethylsilyl)diazomethane (2.0M solution in hexanes)
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Aspiration toxicity Category 1
  • Carcinogenicity Category 1B
  • Flammable liquid Category 2
  • Reproductive toxicity Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity after repeated exposure Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H304-May be fatal if swallowed and enters airways.
  • H350-May cause cancer.
  • H315-Causes skin irritation.
  • H330-Fatal if inhaled.
  • H361f-Suspected of damaging fertility.
  • H373-May cause damage to organs.
  • H411-Toxic to aquatic life with long lasting effects.
  • H336-May cause drowsiness or dizziness.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P331-Do NOT induce vomiting.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'