Learn More
Trimethylsulfonium hydroxide, 0.25M solution in methanol, Thermo Scientific Chemicals
Brand: Thermo Scientific 381360010
42.94 EUR valid until 2024-12-31
Use promo code "21665" to get your promotional price.
View more versions of this product
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Trimethylsulfonium hydroxide | |
0.25M solution in methanol | |
C3H9IS | |
0.2 to 0.3M | |
trimethylsulfonium hydroxide, trimethylsulfonium hydroxide solution, trimethylsulfonium hydroxide 0.2mol/l in methanol methylating reagent, sulfonium, trimethyl-, hydroxide, acmc-209sky, trimethylsulfoniumhydroxyd, ksc498e2f, trimethylsulfoniumhydroxidesolution, trimethylsulfonium hydroxide, 0.2mol/l methanol solution, trimethylsulfoniumhydroxide 0.2mol/linmethanol methylatingreagent | |
[I-].C[S+](C)C | |
204.07 | |
94.18 | |
1 mL |
Colorless | |
17287-03-5, 67-56-1 | |
MFCD00011632 | |
0.81g/mL | |
VFJYIHQDILEQNR-UHFFFAOYSA-M | |
trimethylsulfanium iodide | |
11105313 | |
Glass bottle | |
Liquid |
Safety and Handling
- Trimethylsulfonium hydroxide, 0.25M solution in methanol
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- Serious eye damage/eye irritation Category 1
- Flammable liquid Category 2
- Skin corrosion/irritation Category 1 B
- Specific target organ toxicity after single exposure Category 1
Hazard Statement
- H318-Causes serious eye damage.
- H370-Causes damage to organs.
- H314-Causes severe skin burns and eye damage.
- H301-Toxic if swallowed.
- H311-Toxic in contact with skin.
- H331-Toxic if inhaled.
- H225-Highly flammable liquid and vapour.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P240-Ground/bond container and receiving equipment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
RUO – Research Use Only