missing translation for 'onlineSavingsMsg'
Learn More

Trimethylsulfoxonium iodide, 98%, Thermo Scientific Chemicals

Product Code. 10685762
Click to view available options
Quantity:
100 g
500 g
Packaging:
Plastic bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1774-47-6
Molecular Formula C3H9IOS
Molecular Weight (g/mol) 220.07
MDL Number MFCD00011899
InChI Key BPLKQGGAXWRFOE-UHFFFAOYSA-M
Synonym trimethylsulfoxonium iodide, trimethylsulphoxonium iodide, trimethyloxosulfonium iodide, sulfoxonium, trimethyl-, iodide, trimethyloxosulphonium iodide, trimethyl sulphoxonium ioidie, s,s,s-trimethylsulfoxonium iodide, trimethylsulfoxoniumiodide, sulfonium, trimethyl-, iodide, oxide, sulfoxonium, trimethyl-, iodide 1:1
PubChem CID 74498
IUPAC Name trimethyl(oxo)-λ⁶-sulfanylium iodide
SMILES [I-].C[S+](C)(C)=O

Specifications

Melting Point 175.0°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Plastic bottle
Linear Formula (CH3)3S(O)I
Quantity 500 g
Solubility Information Solubility in water: 15g/L (20°C). Other solubilities: insoluble in ethanol and ether
Formula Weight 220.07
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material Trimethylsulfoxonium iodide, 98%
Show More Show Less
compliance-icons
Product Identifier
  • Trimethylsulfoxonium iodide
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • ACUTE AQUATIC Acute 1
Hazard Statement
  • H400-Very toxic to aquatic life.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P391-Collect spillage.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'