Learn More
Trimethyltin chloride
Description
Trimethyltin chloride is used as a heat stabilizer in the manufacture of vinyl window blinds. It is also used as a stabilizer for plastics and as additive to paint, as antifouling agent. It also serves as a byproduct of methyltin manufacture. Further, it is used as a precursor to vinyltrimethylstannane and in polyvinyl chloride stabilization.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifikationer
| Melting Point | 37°C to 39°C |
| Color | White |
| Boiling Point | 154°C |
| Flash Point | 97°C (207°F) |
| Odor | Unpleasant |
| Quantity | 50 g |
| UN Number | UN3146 |
| Sensitivity | Moisture sensitive |
| Solubility Information | Soluble in water,chloroform and organic solvents. |
| Formula Weight | 199.25 |
| Visa mer |
RUO – Research Use Only
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.