Learn More
Trimethyltin hydroxide, 98%, Thermo Scientific Chemicals
Trimethyltin hydroxide, 98%, C3H11OSn, CAS Number-56-24-6, hydroxytrimethyltin, trimethylstannyl hydroxide, trimethyl tin hydroxide, stannane, hydroxytrimethyl, tin, trimethyl-, hydroxide, trimethyltin hydroxide, hydroxytrimethylstannane, trimethylstannanol, trimethylhydroxytin, unii-10alt8dj2v, 25g
1288.00 €
Chemical Identifiers
CAS | 56-24-6 |
---|---|
Molecular Formula | C3H10OSn |
Molecular Weight (g/mol) | 180.82 |
MDL Number | MFCD00013929 |
InChI Key | OJZNYUDKNVNEMV-UHFFFAOYSA-M |
Synonym | trimethyltin hydroxide, stannane, hydroxytrimethyl, trimethylstannanol, hydroxytrimethyltin, trimethyl tin hydroxide, trimethylhydroxytin, hydroxytrimethylstannane, trimethylstannyl hydroxide, unii-10alt8dj2v, tin, trimethyl-, hydroxide |
PubChem CID | 6328620 |
IUPAC Name | trimethyltin;hydrate |
SMILES | C[Sn](C)(C)O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11354859
|
Thermo Scientific Alfa Aesar
071167.14 |
25 g | N/A | ||||||
Description
Trimethyltin hydroxide is used in the synthesis of modified thymines, which is useful as inhibitors of RNase A. It is also used in the preparation of ketoheterocycle inhibitors of fatty acid amide inhibitors. It reacts with 2,2-disubstituted benzothiazolines to form trialkyltin derivatives of sulphur containing Schiff base complexes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
56-24-6 | |
180.82 | |
OJZNYUDKNVNEMV-UHFFFAOYSA-M | |
6328620 | |
C[Sn](C)(C)O |
C3H10OSn | |
MFCD00013929 | |
trimethyltin hydroxide, stannane, hydroxytrimethyl, trimethylstannanol, hydroxytrimethyltin, trimethyl tin hydroxide, trimethylhydroxytin, hydroxytrimethylstannane, trimethylstannyl hydroxide, unii-10alt8dj2v, tin, trimethyl-, hydroxide | |
trimethyltin;hydrate |
Specifications
56-24-6 | |
White | |
C3H10OSn | |
MFCD00013929 | |
Moisture sensitive | |
Soluble in water. | |
C[Sn](C)(C)O | |
180.82 | |
180.8 | |
Powder |
114°C to 118°C | |
Pungent | |
(CH3)3SnOH | |
UN3146 | |
trimethyltin hydroxide, stannane, hydroxytrimethyl, trimethylstannanol, hydroxytrimethyltin, trimethyl tin hydroxide, trimethylhydroxytin, hydroxytrimethylstannane, trimethylstannyl hydroxide, unii-10alt8dj2v, tin, trimethyl-, hydroxide | |
OJZNYUDKNVNEMV-UHFFFAOYSA-M | |
trimethyltin;hydrate | |
6328620 | |
98% | |
Trimethyltin hydroxide |
Safety and Handling
GHS H Statement
H300-H310-H330
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
P260-P262-P264b-P270-P271-P280-P284-P301+P310-P302+P350-P304+P340-P310-P330-P361-P363-P501c
H300+H310+H330
missing translation for 'dotInformation' : Hazard Class: 6.1; Packaging Group: III
missing translation for 'rtecsNumber' : WH8400000
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures; Store under Nitrogen
- Trimethyltin hydroxide
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 1
- Acute toxicity Category 2
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
Hazard Statement
- H300-Fatal if swallowed.
- H310-Fatal in contact with skin.
- H330-Fatal if inhaled.
- H400-Very toxic to aquatic life.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
- MIXTURE LIST-Contains : Trimethyltin hydroxide
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.