missing translation for 'onlineSavingsMsg'
Learn More

Triphenyl phosphate, 99+%, Thermo Scientific Chemicals

26.60 € - 168.00 €

Chemical Identifiers

CAS 115-86-6
Molecular Formula C18H15O4P
Molecular Weight (g/mol) 326.29
MDL Number MFCD00003031
InChI Key XZZNDPSIHUTMOC-UHFFFAOYSA-N
Synonym triphenylphosphate, phosphoric acid, triphenyl ester, disflamoll tp, triphenoxyphosphine oxide, celluflex tpp, phosflex tpp, trifenylfosfat, phenyl phosphate pho 3po, phosphoric acid triphenyl ester, trifenylfosfat czech
PubChem CID 8289
ChEBI CHEBI:35033
IUPAC Name triphenyl phosphate
SMILES O=P(OC1=CC=CC=C1)(OC1=CC=CC=C1)OC1=CC=CC=C1
View More Specs

Products 4
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10051660
View Documents Promotion Details
Thermo Scientific Acros
147672500
250 g Plastic bottle
26.60 €
250g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
10561581
View Documents Promotion Details
Thermo Scientific Acros
147670010
1 kg Plastic bottle
51.00 €
1kg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
10256333
View Documents Promotion Details
Thermo Scientific Acros
147670025
2.5 kg Plastic bottle
114.00 €
2.50kg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
10752705
View Documents Promotion Details
Thermo Scientific Acros
147670050
5 kg Plastic drum
168.00 €
5kg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Specifications

Chemical Identifiers

115-86-6
326.29
XZZNDPSIHUTMOC-UHFFFAOYSA-N
8289
triphenyl phosphate
C18H15O4P
MFCD00003031
triphenylphosphate, phosphoric acid, triphenyl ester, disflamoll tp, triphenoxyphosphine oxide, celluflex tpp, phosflex tpp, trifenylfosfat, phenyl phosphate pho 3po, phosphoric acid triphenyl ester, trifenylfosfat czech
CHEBI:35033
O=P(OC1=CC=CC=C1)(OC1=CC=CC=C1)OC1=CC=CC=C1

Specifications

0.05mg KOH/g max.
47°C to 53°C
370°C
Authentic
C18H15O4P
MFCD00003031
15, 9915
Solubility in water: insoluble. Other solubilities: soluble in ethanol and acetone
O=P(OC1=CC=CC=C1)(OC1=CC=CC=C1)OC1=CC=CC=C1
326.29
CHEBI:35033
99+%
Triphenyl phosphate
115-86-6
White
220°C
99% min. (GC)
(C6H5O)3P(O)
06, 179
triphenylphosphate, phosphoric acid, triphenyl ester, disflamoll tp, triphenoxyphosphine oxide, celluflex tpp, phosflex tpp, trifenylfosfat, phenyl phosphate pho 3po, phosphoric acid triphenyl ester, trifenylfosfat czech
XZZNDPSIHUTMOC-UHFFFAOYSA-N
triphenyl phosphate
8289
326.28
Crystalline Flakes
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Very toxic to aquatic life with long lasting effects.

GHS P Statement
Avoid release to the environment.
Collect spillage.
Dispose of contents/container to an approved waste disposal plant.

GHS Signal Word: Warning

missing translation for 'einecsNumber' : 204-112-2

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only