missing translation for 'onlineSavingsMsg'
Learn More
Triphenyl phosphite, 99%, Thermo Scientific Chemicals Product Code.: 10163620

Triphenyl phosphite, 99%, Thermo Scientific Chemicals

Product Code. 10163620
1 L, Glass Bottle
Click to view available options
Quantity:
1 L
2.5 L
10 L
Packaging:
Glass Bottle
Plastic drum
This item is not returnable. View return policy

Product Code. 10163620

Brand: Thermo Scientific Acros 140440010

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 101-02-0
Molecular Formula C18H15O3P
Molecular Weight (g/mol) 310.28
InChI Key HVLLSGMXQDNUAL-UHFFFAOYSA-N
Synonym phosphorous acid, triphenyl ester, advance tpp, triphenoxyphosphine, phenyl phosphite, stabilizer p 36, trifenoxyfosfin, trifenylfosfit, phosclere t 36, efed, mellite 310
PubChem CID 7540
IUPAC Name triphenyl phosphite
SMILES C1=CC=C(C=C1)OP(OC2=CC=CC=C2)OC3=CC=CC=C3

Specifications

Acidity 1.5mg KOH/g max.
Melting Point 22.0°C to 24.0°C
Color Colorless to Yellow
Density 1.1840g/mL
Boiling Point 360.0°C
Flash Point 146°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass Bottle
Linear Formula (C6H5O)3P
Refractive Index 1.5893 to 1.5913
Quantity 1 L
Beilstein 06,177
Fieser 01,1249; 02,446; 03,322
Specific Gravity 1.184
Solubility Information Solubility in water: insoluble. Other solubilities: slowly hydrolyses in water, soluble in many organic solvents
Formula Weight 310.28
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Triphenyl phosphite
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0460657 Certificate TypeCertificate of Analysis Date15/03/2024 Catalog Number140440025,140440010,140440100
Lot NumberA0447346 Certificate TypeCertificate of Analysis Date09/11/2023 Catalog Number140440025,140440010,140440100
Lot NumberA0453363 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number140440025,140440010,140440100
Lot NumberA0438215 Certificate TypeCertificate of Analysis Date24/01/2023 Catalog Number140440025,140440010,140440100
Lot NumberA0400133 Certificate TypeCertificate of Analysis Date15/12/2022 Catalog Number140440025,140440010,140440100
compliance-icons compliance-icons
Product Identifier
  • Triphenyl phosphite
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
  • Serious eye damage/eye irritation Category 2
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
  • H319-Causes serious eye irritation.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/and soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P273-Avoid release to the environment.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Triphenyl phosphite, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.