Learn More
Description
Triphenylene is used in optics and electronics. It is also used as a discotic mesogen in liquid crystalline materials. Further, it is a compound that fluoresces in the ultraviolet region. In addition to this, it is used in the preparation of triphenylene-2-carbaldehyde.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Triphenylene is used in optics and electronics. It is also used as a discotic mesogen in liquid crystalline materials. Further, it is a compound that fluoresces in the ultraviolet region. In addition to this, it is used in the preparation of triphenylene-2-carbaldehyde.
Solubility
Soluble in ethanol, benzene, acetic acid, oils and chloroform.
Notes
Incompatible with strong oxidizing agents.

Specifications
Specifications
Melting Point | 194°C to 199°C |
Density | 1.302 |
Boiling Point | 438°C |
Quantity | 5 g |
Beilstein | 1342908 |
Merck Index | 14,9740 |
Solubility Information | Soluble in ethanol,benzene,acetic acid,oils and chloroform. |
Formula Weight | 228.3 |
Percent Purity | 98% |
Chemical Name or Material | Triphenylene |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
10159323 | Certificate of Analysis | 08/12/2021 | L00955.03,L00955.06 |
10127018 | Certificate of Analysis | 08/12/2021 | L00955.03,L00955.06 |
10143322 | Certificate of Analysis | 08/12/2021 | L00955.03,L00955.06 |
M21F006 | Certificate of Analysis | 08/12/2021 | L00955.03,L00955.06 |
10148940 | Certificate of Analysis | 07/12/2021 | L00955.03,L00955.06 |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.