missing translation for 'onlineSavingsMsg'
Learn More

Triphenylmethane, 99+%

Product Code. 10041670 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
25g
500g
This item is not returnable. View return policy

Product Code. 10041670

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 519-73-3 | C19H16 | 244.34 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 519-73-3
Molecular Formula C19H16
Molecular Weight (g/mol) 244.34
MDL Number MFCD00004763
InChI Key AAAQKTZKLRYKHR-UHFFFAOYSA-N
Synonym triphenylmethane, tritane, benzene, 1,1',1-methylidynetris, methane, triphenyl, diphenylmethyl benzene, 1,1',1-methanetriyltribenzene, unii-8o4utw9e17, ccris 5194, triphenyl methane, triphenylmethane?
PubChem CID 10614
ChEBI CHEBI:76212
IUPAC Name benzhydrylbenzene
SMILES C1=CC=C(C=C1)C(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 90.0°C to 94.0°C
Color Yellow
Density 1.0100g/mL
Boiling Point 358.0°C to 359.0°C (754.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Linear Formula (C6H5)3CH
Quantity 25 g
Beilstein 05,698
Merck Index 15,9914
Specific Gravity 1.01
Solubility Information Solubility in water: insoluble in water
Formula Weight 244.34
Percent Purity 99+%
Physical Form Powder
Chemical Name or Material Triphenylmethane, 99+%
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.