missing translation for 'onlineSavingsMsg'
Learn More

Triphenylphosphine resin, 1% crosslinked with DVB, 1.0-1.5mmol/g, 100-200 mesh, Thermo Scientific Chemicals

Product Code. 10025403
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 39319-11-4
Molecular Formula C18H15P
Molecular Weight (g/mol) 262.29
MDL Number MFCD00003043 MFCD20489348
InChI Key RIOQSEWOXXDEQQ-UHFFFAOYSA-N
PubChem CID 11776
IUPAC Name triphenylphosphane
SMILES C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Physical Form 100 To 200 Mesh
Packaging Glass bottle
Quantity 25 g
Chemical Name or Material Triphenylphosphine resin, 1% crosslinked with DVB, 1.0-1.5mmol/g

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'