missing translation for 'onlineSavingsMsg'
Learn More

Triphenylsilane, 97%, Thermo Scientific Chemicals

Product Code. 11414693
Click to view available options
Quantity:
10 g
50 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Triphenylsilane acts as a reactant or reagent for catalytic hydrogen deuterium exchange reactions of silanes, to be oxidized by carbon nanotube-gold nanohybrids, for hydrolysis by ruthenium complexes, for hydrosilylation to produce enolsilanes, for synthesis of bromosilanes, for ozone oxidation of silyl-alkenes for synthesis of α-O-silylated acyloin derivatives. It is used to synthesize hydrido silyl and bis(silyl) bis(imidazolinylidene) nickel complexes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Triphenylsilane acts as a reactant or reagent for catalytic hydrogen deuterium exchange reactions of silanes, to be oxidized by carbon nanotube-gold nanohybrids, for hydrolysis by ruthenium complexes, for hydrosilylation to produce enolsilanes, for synthesis of bromosilanes, for ozone oxidation of silyl-alkenes for synthesis of α-O-silylated acyloin derivatives. It is used to synthesize hydrido silyl and bis(silyl) bis(imidazolinylidene) nickel complexes.

Solubility
Soluble in methanol. Decomposes in water.

Notes
Air Sensitive. Incompatible with air and oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemische Identifikatoren

CAS 789-25-3
Molecular Formula C18H15Si
Molecular Weight (g/mol) 259.403
MDL Number MFCD00003003
InChI Key BZLZKLMROPIZSR-UHFFFAOYSA-N
Synonym triphenylsilane, benzene, 1,1',1-silylidynetris, triphenylsilyl, tri phenyl silicon, triphenylsilyl radical, triphenylsilane 5g, 1,1 inverted exclamation marka,1 inverted exclamation marka inverted exclamation marka-silylidynetrisbenzene; nsc 12565; triphenylhydrosilane
PubChem CID 6327682
IUPAC Name triphenylsilicon
SMILES C1=CC=C(C=C1)[Si](C2=CC=CC=C2)C3=CC=CC=C3

Spezifikation

Boiling Point 159°C to 163°C (3mmHg)
Melting Point 42°C to 44°C
Quantity 10 g
Beilstein 978182
Flash Point 76°C (169°F)
Solubility Information Soluble in methanol. Decomposes in water.
Formula Weight 260.41
Percent Purity 99%
Sensitivity Moisture sensitive
Chemical Name or Material Triphenylsilane
compliance-icons
Produktidentifikator
  • Triphenylsilane
Signalwort
  • Warning
Gefahrenkategorie
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Gefahrenhinweis
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Sicherheitshinweis
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Ergänzende Informationen
  • MIXTURE LIST-Contains : Triphenylsilane

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'