missing translation for 'onlineSavingsMsg'
Learn More

Triphenyltin hydroxide, Thermo Scientific™

44.20 € - 144.00 €

Chemical Identifiers

CAS 76-87-9
Molecular Formula C18H18OSn
Molecular Weight (g/mol) 369.05
MDL Number MFCD00013928
InChI Key ZJIGGMIMCKZRRB-UHFFFAOYSA-N
Synonym triphenyltin hydroxide, fentin hydroxide, triphenylstannanol, vancide ks, hydroxytriphenyltin, hydroxytriphenylstannane, erithane, fenolovo, tenhide, stannane, hydroxytriphenyl
PubChem CID 6327657
IUPAC Name triphenyltin;hydrate
SMILES O.C1=CC=C(C=C1)[SnH](C1=CC=CC=C1)C1=CC=CC=C1
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11384859
View Documents
Thermo Scientific Alfa Aesar
071177.14
25 g
44.20 €
25g
Please to purchase this item. Need a web account? Register with us today!
11394859
View Documents
Thermo Scientific Alfa Aesar
071177.22
100 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

76-87-9
369.05
ZJIGGMIMCKZRRB-UHFFFAOYSA-N
6327657
O.C1=CC=C(C=C1)[SnH](C1=CC=CC=C1)C1=CC=CC=C1
C18H18OSn
MFCD00013928
triphenyltin hydroxide, fentin hydroxide, triphenylstannanol, vancide ks, hydroxytriphenyltin, hydroxytriphenylstannane, erithane, fenolovo, tenhide, stannane, hydroxytriphenyl
triphenyltin;hydrate

Specifications

76-87-9
White
(C6H5)3SnOH
UN3146
triphenyltin hydroxide, fentin hydroxide, triphenylstannanol, vancide ks, hydroxytriphenyltin, hydroxytriphenylstannane, erithane, fenolovo, tenhide, stannane, hydroxytriphenyl
ZJIGGMIMCKZRRB-UHFFFAOYSA-N
triphenyltin;hydrate
6327657
Powder
122°C to 124°C
C18H18OSn
MFCD00013928
14,9745
Slightly soluble in alcohol, toluene
O.C1=CC=C(C=C1)[SnH](C1=CC=CC=C1)C1=CC=CC=C1
369.05
367.01
Triphenyltin hydroxide
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H311-H330-H372-H351-H361-H318-H315-H335
Toxic if swallowed.
Toxic in contact with skin.
Fatal if inhaled.
Causes damage to organs through prolonged or repeated exposure.
Suspected of causing cancer.
Suspected of damaging fertility or the unborn child.
Causes serious eye damage.
Causes skin irritation.
May cause respiratory irritation.

P201-P202-P260-P264b-P270-P271-P281-P284-P301+P310-P302+P352-P304+P340-P305+P351+P338-P308+P313-P310-P312-P330-P332+P313-P361-P363-P501c

H301+H311-H315-H318-H330-H335-H351-H361d-H372

missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ORGANOTIN COMPOUNDS, SOLID, N.O.S.

missing translation for 'einecsNumber' : 200-990-6

missing translation for 'rtecsNumber' : WH8575000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.