missing translation for 'onlineSavingsMsg'
Learn More

Triphenylvinylsilane, 98%, Thermo Scientific Chemicals

Product Code. 11410643
Click to view available options
Quantity:
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Triphenylvinylsilane can be used to produce triphenyl-[2-(tetrahydro-furan-2-yl)-ethyl]-silane at temperature of 190°C. This reaction will need reagent di(t-butyl)peroxide with reaction time of 2 hours. Also employed as an organic light emitting diode.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Triphenylvinylsilane can be used to produce triphenyl-[2-(tetrahydro-furan-2-yl)-ethyl]-silane at temperature of 190°C. This reaction will need reagent di(t-butyl)peroxide with reaction time of 2 hours. Also employed as an organic light emitting diode.

Solubility
Insoluble in water.

Notes
Moisture Sensitive. Store away from oxidizing agents, water/moisture and heat. Store under inert gas.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18666-68-7
Molecular Formula C20H18Si
Molecular Weight (g/mol) 286.449
MDL Number MFCD00051542
InChI Key OVOIHGSHJGMSMZ-UHFFFAOYSA-N
Synonym triphenylvinylsilane, silane, ethenyltriphenyl, triphenyl vinyl silane, silane, triphenylvinyl, vinyltriphenylsilane, benzene, 1,1',1-ethenylsilylidyne tris, ethenyltriphenylsilan, vinyltriphenyl silane, vinyl triphenyl silane, ethenyl triphenyl silane
PubChem CID 87747
IUPAC Name ethenyl(triphenyl)silane
SMILES C=C[Si](C1=CC=CC=C1)(C2=CC=CC=C2)C3=CC=CC=C3

Specifications

Melting Point 68°C to 71°C
Quantity 5 g
Linear Formula (C6H5)3SiCH=CH2
Beilstein 913423
Solubility Information Insoluble in water.
Formula Weight 286.45
Percent Purity 98%
Sensitivity Moisture sensitive
Chemical Name or Material Triphenylvinylsilane

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.