missing translation for 'onlineSavingsMsg'
Learn More
Triphosgene, 98%, Thermo Scientific Chemicals Product Code.: 11452757

Triphosgene, 98%, Thermo Scientific Chemicals

Code produit. 11452757
100 g
missing translation for 'orderingAttributeHoverText'
Quantity:
5 g
25 g
100 g
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

Product Code. 11452757

Brand: Thermo Scientific Alfa Aesar A14932.22

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

Triphosgene is used as a carbonylating agent for aza-peptide synthesis. It reacts with several alfa-amino acids to give the corresponding N-carboxyanhydrides. It is involved in the preparation of the esterification coupling reagent, di-2-thienyl carbonate from 2(5H)-thiophenone. Further, it is used as a reagent in organic synthesis and converts an amino group into isocyanate. In addition to this, it is employed in the preparation of 2-chloronicotinaldehydes through cyclization of the corresponding enamides. It is considered as a useful substitute for phosgene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Triphosgene is used as a carbonylating agent for aza-peptide synthesis. It reacts with several alfa-amino acids to give the corresponding N-carboxyanhydrides. It is involved in the preparation of the esterification coupling reagent, di-2-thienyl carbonate from 2(5H)-thiophenone. Further, it is used as a reagent in organic synthesis and converts an amino group into isocyanate. In addition to this, it is employed in the preparation of 2-chloronicotinaldehydes through cyclization of the corresponding enamides. It is considered as a useful substitute for phosgene.

Solubility
Soluble in ether, ethanol, tetrahydrofuran, benzene, cyclohexane, chloroform, hexane, carbon tetrachloride, dichloromethane and chlorobenzene. Insoluble in water.

Notes
Moisture sensitive. Store in a cool place. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Identifiants chimiques

CAS 32315-10-9
Molecular Formula C3Cl6O3
Molecular Weight (g/mol) 296.73
MDL Number MFCD00062848
InChI Key UCPYLLCMEDAXFR-UHFFFAOYSA-N
Synonym triphosgene, bis trichloromethyl carbonate, methanol, trichloro-, carbonate 2:1, ditrichloromethyl carbonate, triphosgene bis-trichloromethyl carbonate, methanol, 1,1,1-trichloro-, 1,1'-carbonate, triphosgen, tri phosgene, tri-phosgene
PubChem CID 94429
IUPAC Name bis(trichloromethyl) carbonate
SMILES C(=O)(OC(Cl)(Cl)Cl)OC(Cl)(Cl)Cl

Spécification

Melting Point 77°C to 91°C
Density 1.6
Boiling Point 203°C to 206°C
Quantity 100 g
UN Number UN2928
Beilstein 1787583
Sensitivity Moisture sensitive
Solubility Information Soluble in ether,ethanol,tetrahydrofuran,benzene,cyclohexane,chloroform,hexane,carbon tetrachloride,dichloromethane and chlorobenzene. Insoluble in water.
Formula Weight 296.75
Percent Purity 98%
Chemical Name or Material Triphosgene
Afficher plus Afficher moins
compliance-icons compliance-icons
Identifiant produit
  • Triphosgene
Mention d'avertissement
  • Danger
Catégorie de danger
  • Acute toxicity Category 2
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Mention de danger
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H330-Fatal if inhaled.
Conseils de prudence
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Informations supplémentaires
  • MIXTURE LIST-Contains : Triphosgene

RUO – Research Use Only

Correction du contenu d'un produit

Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.

Nom du produit
Triphosgene, 98%, Thermo Scientific Chemicals >

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.

Merci de nous aider à améliorer notre site web. Votre commentaire a été soumis