Learn More
Fisherbrand™ Triple Instrument Bundle
Whether you need to spin-down, shake, or stir, this Triple Instrument Bundle has you covered.
Brand:  Fisherbrand™ 120950
Product Code. 17862966
  • 1050.00 € / 1 set
Edge
Explore more special offers
Description
Description
  • Get the full functionality of three machines in two pieces of equipment! Complete with a Fisherbrand Silicone Lab Mat to protect your workspace from spills and wear.
  • Fisherbrand Centrifuge/Stirrer: The industry's first high-speed centrifuge (12,500 rpm) and magnetic stirrer (3 L), combined into one unit. Made durable and chemically resistant with easy cleaning and maintenance-free operation. Its compact and low-profile design optimizes limited bench space and minimizes clutter in the lab.
  • Fisherbrand Mini Vortexer: Small footprint Vortexer offering maximum vortex shaking for a variety of tube sizes. A simple touch start operation allows for immediate sample mixing with one hand and automatic stop when tube is lifted.
  • Fisherbrand Silicone Lab Mat: This FDA food-grade silicone labmat is suitable for various laboratory settings, including cold rooms, clean rooms, and sterile suites, where high sterility is essential. Effectively safeguards countertops from temperatures up to 200°C (392°F) and demonstrates general chemical resistance to moderate oxidizing chemicals.
Specifications
Specifications
100/240 V
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Fisherbrand™ Triple Instrument Bundle > Fisherbrand Triple Instrument