missing translation for 'onlineSavingsMsg'
Learn More

H-B DURAC™ SP Scienceware™ Triple Scale Beer and Wine Hydrometers

DURAC Triple Scale Beer and Wine Hydrometers allows for optimal beer brewing and wine making. Includes three scales, Specific Gravity, Potential Alcohol by Volume (PAV), and Brix, that provide invaluable data points when crafting beer, wine, kombucha, mead, cider, and more in any setting.

Brand:  H-B DURAC™ B61800-6100


 View more versions of this product

Product Code. 17460244

  • 35.35 € / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Determine if starting mash or wort is acceptable quality to achieve desired results
  • Monitor fermentation process to see if variables should be altered
  • Determine when fermentation is complete
  • Calculate approximate alcohol content of the finished product; from 0 to 20% by volume
  • Manufactured in strict accordance with NIST as outlined in NBS Circular 555 Military STD-45662A and ISO 17025 where applicable.
  • Meets or exceeds the tolerances of accuracy and physical specifications set forth by the NIST Circular 55, American Society of testing Materials e100-10 or the American Petroleum Institute
  • Lead and mercury free

Available in 3 styles:

  • Shatterproof
    • All plastic, shatterproof design removes risk of contamination due to instrument breakage
    • Easy to read flat surface design
  • Polycarbonate
    • Optical quality polycarbonate provides glass-like clarity with shatterproof strength to eliminate the risk of contamination due to instrument breakage
    • Color coded bands indicate when fermentation is complete; easy to read font
  • Glass
    • Standard glass design provides inexpensive fermentation monitoring
    • Color coded bands indicate when fermentation is complete; easy to read font

 

Order Info

Recommend testing in a Hydrometer Jar (sold separately).

Services

1 Year Warranty

Specifications

Specifications

1
±0.005 SpGr; 1° Brix
NIST
Hydrometer jar; Thermometer
0 to 35°
Beer brewing; Wine making
20°C
Alcohol Hydrometer
0.005 sp.gr.; 1° Brix; ABV: 1%
234 mm
Conversion chart
Steel Shot
1.00 to 1.15
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • H-B DURAC TRIPLE SCALE BEER AND WINE HYDROMETER, SHATTERPROOF

Signal Word
  • Warning

Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1

Hazard Statement
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
  • H319-Causes serious eye irritation.
  • H411-Toxic to aquatic life with long lasting effects.

Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : Reaction products ofEpichlorohydrin andBisphenol A

SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

All scales are meant to be read with liquids at 68°F (20°C). A conversion chart is included to adjust for varying temperatures. For accurate results, the hydrometer should always be used in conjunction with a thermometer (sold separately).