Learn More
Thermo Scientific™ TriPlus RSH Autosampler Fixed Needle Syringe, 10μL
Thermo Scientific™ TriPlus RSH Autosampler Fixed Needle Syringes are accurate and reproducible over a range of volumes.
78.39 € - 133.17 €
Specifications
Volume (Metric) 10 μL
Needle Point Style Cone
View More Specs
Products 2
Product Code Brand Needle Gauge Length (Metric) Needle Price Quantity & Availability  
Product Code Brand Needle Gauge Length (Metric) Needle Price Quantity & Availability  
13406856
View Documents
Thermo Scientific™
365D0271
23s G 57 mm
78.39 €
Each
13436856
View Documents
Thermo Scientific™
365D0321
26s G 85 mm
133.17 €
Each
Estimated Shipment: 04-02-2025
to see stock.
Add to basket
Description
Description
Thermo Scientific™ TriPlus RSH Autosampler Fixed Needle Syringes are accurate and reproducible over a range of volumes.
  • Borosilicate glass with metal flanges
  • Needles and plungers are stainless steel
  • Field-proven syringe design
Specifications
Specifications
10 μL
Cone