missing translation for 'onlineSavingsMsg'
Learn More

Tripropionin, 97%, Thermo Scientific Chemicals

Product Code. 10551311
Click to view available options
Quantity:
100 g
250 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 139-45-7
Molecular Formula C12H20O6
Molecular Weight (g/mol) 260.28
MDL Number MFCD00027005
InChI Key YZWRNSARCRTXDS-UHFFFAOYSA-N
Synonym tripropionin, glycerol tripropionate, glyceryl tripropionate, tripropionine, glyceryl tripropanoate, propionin, tri, glycerine tripropionate, tripropionylglycerol, 1,2,3-propanetriol, tripropanoate, propionic acid triglyceride
PubChem CID 8763
IUPAC Name 2,3-di(propanoyloxy)propyl propanoate
SMILES CCC(=O)OCC(COC(=O)CC)OC(=O)CC

Specifications

Color Colorless
Density 1.0820g/mL
Boiling Point 151°C (2.0 mmHg)
Flash Point 162°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.4310 to 1.434
Quantity 100 g
Specific Gravity 1.082
Formula Weight 260.28
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Tripropionin, 97%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'