missing translation for 'onlineSavingsMsg'
Learn More

Tris[(1-benzyl-1H-1,2,3-triazol-4-yl)methyl]amine, 97+%, Thermo Scientific Chemicals

Product Code. 15420107
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Tris[(1-benzyl-1H-1,2,3-triazol-4-yl)methyl]amine stabilizes Cu(I) towards disproportionation and oxidation thus enhancing its catalytic effect in the azide-acetylene cycloaddition.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tris[(1-benzyl-1H-1,2,3-triazol-4-yl)methyl]amine stabilizes Cu(I) towards disproportionation and oxidation thus enhancing its catalytic effect in the azide-acetylene cycloaddition.

Solubility
Soluble in DMSO, DMF.

Notes
Store at -20°C. Protect from heat. Incompatible with heat and oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 510758-28-8
Molecular Formula C30H30N10
Molecular Weight (g/mol) 530.64
MDL Number MFCD09265124
InChI Key WKGZJBVXZWCZQC-UHFFFAOYSA-N
Synonym tris 1-benzyl-1h-1,2,3-triazol-4-yl methyl amine, 1,2,4-trimethyl-5-benzenesulfonic acid, tris 1-benzyl-1h-1,2,3-triazole-4-ylmethyl amine, tris 1-benzyl-1h-1,2,3-triazol-4-yl methyl amine tbta
PubChem CID 11203363
IUPAC Name 1-(1-benzyltriazol-4-yl)-N,N-bis[(1-benzyltriazol-4-yl)methyl]methanamine
SMILES C(N(CC1=CN(CC2=CC=CC=C2)N=N1)CC1=CN(CC2=CC=CC=C2)N=N1)C1=CN(CC2=CC=CC=C2)N=N1

Specifications

Melting Point 132°C to 143°C
Color White
Quantity 250 mg
Solubility Information Soluble in DMSO,DMF.
Formula Weight 530.63
Percent Purity ≥97%
Physical Form Solid
Chemical Name or Material Tris[(1-benzyl-1H-1,2,3-triazol-4-yl)methyl]amine
compliance-icons
Product Identifier
  • Tris[(1-benzyl-1H-1,2,3-triazol-4-yl)methyl]amine
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : Tris[(1-benzyl-1H-1,2,3-triazol-4-yl)methyl]amine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.