missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Tris(2-carboxyethyl)phosphine hydrochloride, 98%, Molecular Biology Grade

Product Code. 15450127
Click to view available options
Quantity:
2 g
5 g
10 g

For reducing disulfide bonds in peptides and proteins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 51805-45-9
Molecular Formula C9H12O6P
Molecular Weight (g/mol) 247.16
MDL Number MFCD00145469
InChI Key PZBFGYYEXUXCOF-UHFFFAOYSA-K
Synonym tris 2-carboxyethyl phosphine hydrochloride, tcep hcl, tcep hydrochloride, 3,3',3-phosphinetriyltripropanoic acid hydrochloride, tcep, unii-h49aam893k, tris carboxyethyl phosphine hydrochloride, tris-2-carboxyethyl phosphine hydrochloride, tris 2-carboxyethyl phosphine hcl, 3,3',3-phosphinetriyltripropanoicacidhydrochloride
PubChem CID 2734570
IUPAC Name 3-[bis(2-carboxyethyl)phosphanyl]propanoic acid;hydrochloride
SMILES [O-]C(=O)CCP(CCC([O-])=O)CCC([O-])=O

Specifications

Melting Point 177°C to 178°C
Packaging Plastic bottle
Quantity 2 g
UN Number UN3261
Sensitivity Moisture sensitive
Solubility Information Soluble in DMSO or water
Formula Weight 286.65
Percent Purity 98%
Grade Molecular Biology
Physical Form Crystalline Powder
Chemical Name or Material Tris(2-carboxyethyl)phosphine hydrochloride
Show More Show Less
compliance-icons
Product Identifier
  • Tris(2-carboxyethyl)phosphine hydrochloride, Molecular Biology Grade
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : Tris(2-carboxyethyl)phosphine hydrochloride

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'