missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tris(8-hydroxyquinolinato)aluminum, 99%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
25 g
Description
Key green-light emitting and electron transport material for OLEDs.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Key green-light emitting and electron transport material for OLEDs.
Solubility
Solubility in chloroform
Notes
Reacts with strong oxidizing agents. Store at room temperature.
Key green-light emitting and electron transport material for OLEDs.
Solubility
Solubility in chloroform
Notes
Reacts with strong oxidizing agents. Store at room temperature.
Specifications
Specifications
Melting Point | >300°C |
Color | Yellow to Green |
Quantity | 5 g |
Merck Index | 14,370 |
Formula Weight | 459.44 |
Percent Purity | 99% |
Physical Form | Powder |
Chemical Name or Material | Tris(8-hydroxyquinolinato)aluminum |
Safety and Handling
Product Identifier
- Tris(8-hydroxyquinolinato)aluminum
- Warning
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- MIXTURE LIST-Contains : Aluminium tris(quinolin-8-olate)
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'