missing translation for 'onlineSavingsMsg'
Learn More

TAE Buffer, Tris-Acetate-EDTA, 10X Solution, Electrophoresis, Fisher BioReagents

29.80 € - 350.00 €

Specifications

Chemical Name or Material Tris-Acetate-EDTA
Name Note 10X Solution
CAS 64-19-7, 7732-18-5, 60-00-4, 77-86-1
Physical Form Liquid
DNase DNase free
View More Specs

Products 4
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10628403
View Documents
Fisher BioReagents
BP1335-500
500 mL Poly Bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10041223
View Documents
Fisher BioReagents
BP1335-1
1 L Nalgene Poly Bottle
40.50 €
1 L
Please to purchase this item. Need a web account? Register with us today!
10775494
View Documents
Fisher BioReagents
BP1335-4
4 L Poly CUBE
108.00 €
4 L
Please to purchase this item. Need a web account? Register with us today!
10637633
View Documents
Fisher BioReagents
BP1335-20
20 L Poly CUBE This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

Tris–Acetate–EDTA (TAE) is commonly used as a buffer for nucleic acid electrophoresis.
Specifications

Specifications

Tris-Acetate-EDTA
64-19-7, 7732-18-5, 60-00-4, 77-86-1
DNase free
DNase-, RNase- and Protease-Free
TAE
Tris (<5.0%), Acetic Acid (1.0%), and EDTA (<1%).
10X Solution
Liquid
Filtered through a 0.2-micron filter.
Protease free
Electrophoresis
Videos
Safety and Handling

Safety and Handling

Recommended Storage : RT

SDS
Documents

Documents

Product Certifications