missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tris Buffer, 2M Solution (Molecular Biology), Fisher BioReagents
Click to view available options
Quantity:
100 mL
500 mL
Packaging:
Poly Bottle
Description
Tris is a buffer component in molecular biology, tissue culture, and electrophoresis procedures. Tris Solution, 2M, provides a convenient stock solution for preparing Tris buffers.
Specifications
Specifications
Color | Undesignated |
Concentration | 1.95 to 2.05 M |
Physical Form | Liquid |
Quantity | 100 mL |
DNase | DNase free |
Identification | Pass Test |
Packaging | Poly Bottle |
Protease | Protease free |
Merck Index | 15, 9951 |
Grade | Molecular Biology |
Show More |
Safety and Handling
Product Identifier
- Tris buffer solution 2M
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction