missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals TRIS-buffered saline (TBS, 10X) pH 7.6

104.52 € - 159.80 €

Chemical Identifiers

CAS 77-86-1
Molecular Formula C4H11NO3
Molecular Weight (g/mol) 121.136
MDL Number MFCD00004679
InChI Key LENZDBCJOHFCAS-UHFFFAOYSA-N
PubChem CID 6503
ChEBI CHEBI:9754
IUPAC Name 2-amino-2-(hydroxymethyl)propane-1,3-diol
SMILES C(C(CO)(CO)N)O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15454989
View Documents Promotion Details
Thermo Scientific Chemicals
J62662.K3
2 L
104.52 €
2 L
Please to purchase this item. Need a web account? Register with us today!
15464989
View Documents Promotion Details
Thermo Scientific Chemicals
J62662.K7
4 L
159.80 €
4 L
Please to purchase this item. Need a web account? Register with us today!
Description

Description

TRIS-buffered saline (TBS, 10X) pH 7.6 is used as a substance diluent, as a wash buffer in immunoassays such as enzyme-linked immunosorbent assay. It is used in antibody diluent in western blotting, sample diluent in vitro diagnostics, immuno-histochemical staining and in situ hybridization.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Miscible with water.

Notes
250mM Tris, 27mM potassium chloride, 1.37M sodium chloride, pH 7.6.Store in a cool place. Incompatible with strong oxidizing agents.
Specifications

Chemical Identifiers

77-86-1
121.136
LENZDBCJOHFCAS-UHFFFAOYSA-N
CHEBI:9754
C(C(CO)(CO)N)O
C4H11NO3
MFCD00004679
6503
2-amino-2-(hydroxymethyl)propane-1,3-diol

Specifications

77-86-1
7.6
C4H11NO3
Miscible with water.
C(C(CO)(CO)N)O
121.136
CHEBI:9754
10X
Liquid
MFCD00004679
LENZDBCJOHFCAS-UHFFFAOYSA-N
2-amino-2-(hydroxymethyl)propane-1,3-diol
6503
TRIS-buffered saline
Videos
Safety and Handling

Safety and Handling

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only