missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals TRIS-buffered saline (TBS, 20X), with 2% Tween 20

124.62 € - 221.10 €

Specifications

Product Type TBS
Form Liquid
Concentration 20X
Content And Storage Ambient temperatures
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15493269
View Documents
Thermo Scientific Chemicals
J60497.K2
1 L
124.62 €
1 L
Please to purchase this item. Need a web account? Register with us today!
15403279
View Documents
Thermo Scientific Chemicals
J60497.K3
2 L
221.10 €
2 L
Please to purchase this item. Need a web account? Register with us today!
Description

Description

TRIS-buffered saline (TBS, 20X), with 2% Tween 20 is used as a wash buffer and for sample preparation in general immunoassay applications, Tween 20 reduces nonspecific binding and protein:protein interactions.TBS is often used for diluting secondary antibodies or streptavidin-AP conjugate. TBS-Tween 20 is often used for washing steps.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
TRIS-buffered saline (TBS, 20X), with 2% Tween 20 is used as a wash buffer and for sample preparation in general immunoassay applications, Tween 20 reduces nonspecific binding and protein:protein interactions.TBS is often used for diluting secondary antibodies or streptavidin-AP conjugate. TBS-Tween 20 is often used for washing steps.

Solubility
It is soluble in water.

Notes
1.0M Tris-HCl, 3.0M NaCl, pH 7.4 ± 0.1 with 2.0% Tween-20
Specifications

Specifications

TBS
Liquid
20X
Ambient temperatures
Videos
SDS
Documents

Documents

Product Certifications

RUO – Research Use Only