missing translation for 'onlineSavingsMsg'
Learn More

Tris(dibenzylideneacetone)dipalladium(0), 97%, Thermo Scientific Chemicals

55.07 € - 910.53 €

Chemical Identifiers

CAS 51364-51-3
Molecular Formula C51H42O3Pd2
Molecular Weight (g/mol) 915.73
MDL Number MFCD00013310
InChI Key CYPYTURSJDMMMP-UHFFFAOYSA-N
Synonym tris dibenzylideneacetone dipalladium 0, tris dibenzylideneacetone dipalladium, pd2 dba 3, tris dibezylideneacetone dipalladium, tris dibenzylideneacetone dipalladium o, tris dibenzylideneacetonyl bis-palladium, tris dba, tris 1e,4e-1,5-diphenylpenta-1,4-dien-3-one dipalladiumShow MoreShow Less
PubChem CID 9811564
IUPAC Name (1E,4E)-1,5-diphenylpenta-1,4-dien-3-one;palladium
SMILES [Pd].[Pd].O=C(C=CC1=CC=CC=C1)C=CC1=CC=CC=C1.O=C(C=CC1=CC=CC=C1)C=CC1=CC=CC=C1.O=C(C=CC1=CC=CC=C1)C=CC1=CC=CC=C1Show MoreShow Less
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10155760
SDS View Documents
Thermo Scientific Acros
318775000
500 mg Glass bottle
55.07 €
500mg
Please to purchase this item. Need a web account? Register with us today!
10170292
SDS View Documents
Thermo Scientific Acros
318770050
5 g Glass bottle
255.27 €
5g
Please to purchase this item. Need a web account? Register with us today!
11954361
SDS View Documents
Thermo Scientific Acros
318770250
25 g Glass bottle
910.53 €
25g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Show More
Videos
Safety and Handling
Show More
SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Tris(dibenzylideneacetone)dipalladium(0), 97%, Thermo Scientific Chemicals

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.