missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TE Buffer, Tris-EDTA, 1X Solution, pH 8.0, Molecular Biology, Fisher BioReagents
Brand: Fisher BioReagents BP2473-500
This item is not returnable.
View return policy
Description
- Save time and simplify your buffer preparation step by using ready-made Fisher BioReagents 1X TBE buffer solution
- Eliminates the hassle of dilution or waiting for powder to dissolve
- 1X solution contains 10mM tris and 1mM EDTA
- Filtered and autoclaved - ready to use
- Tested for the absence of DNase, RNase, and protease
- TE buffer (pH 8.0) is routinely used for resuspending DNA after precipitation
- Intended for laboratory research use only

Specifications
Tris-EDTA | |
77-86-1, 6381-92-6, 7647-01-0, 7732-18-5 | |
7.4 to 8.1 | |
500 mL | |
Filtered through a 5-micron filter. | |
Protease free | |
Molecular Biology |
1X Solution, pH 8.0 | |
Colorless | |
Liquid | |
DNase free | |
Poly Bottle | |
TE |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
TE Buffer, Tris-EDTA, 1X Solution, pH 8.0, Molecular Biology, Fisher BioReagents > Poly Bottle; 500mL
Spot an opportunity for improvement?Share a Content Correction