Learn More
TE Buffer, Tris-EDTA, 10X Solution, pH 7.4, Molecular Biology, Fisher BioReagents
21.35 € - 42.80 €
Specifications
Chemical Name or Material | Tris-EDTA (TE), 10X Solution |
---|---|
Name Note | pH 7.4 |
CAS | 77-86-1, 6381-92-6, 7647-01-0, 7732-18-5 |
Color | Colorless |
pH | 7.4 to 8.1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
11432964
|
Fisher BioReagents
BP2477-100 |
100 mL | Poly Bottle | N/A | |||||
10489693
|
Fisher BioReagents
BP2477-500 |
500 mL | Poly Bottle | N/A | |||||
10387033
|
Fisher BioReagents
BP2477-1 |
1 L | Nalgene Poly Bottle | N/A | |||||
Description
Tris–EDTA (TE) is used for suspending nucleic acid samples.Specifications
Tris-EDTA (TE), 10X Solution | |
77-86-1, 6381-92-6, 7647-01-0, 7732-18-5 | |
7.4 to 8.1 | |
DNase free | |
Protease free | |
Molecular Biology |
pH 7.4 | |
Colorless | |
Liquid | |
Filtered through a 5-micron filter and autoclaved. | |
TE |
Safety and Handling
CAUTION!
Emergency Overview
May cause eye, skin, and respiratory tract irritation. Avoid contact with skin and eyes. Do not breathe dust. Do not breathe vapors or spray mist. Wash off immediately with soap and plenty of water removing all contaminated clothes and shoes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Remove from exposure, lie down. Move to fresh air. If breathing is difficult, give oxygen. If not breathing, give artificial respiration. If not breathing, give artificial respiration. Obtain medical attention.
NFPA
Health:1
Flammability:0
Instability:0
Recommended Storage : RT
Your input is important to us. Please complete this form to provide feedback related to the content on this product.