missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tris-Glycine-SDS, 10X Solution (Electrophoresis), Fisher BioReagents™
50.70 € - 115.00 €
Specifications
Chemical Name or Material | Tris-Glycine-SDS |
---|---|
Name Note | 10X Solution |
CAS | 77-86-1, 7732-18-5, 56-40-6, 151-21-3 |
Color | Undesignated |
Physical Form | Liquid |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10051653
|
Fisher BioReagents
BP1341-1 |
1 L | Nalgene Poly Bottle |
50.70 €
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10102823
|
Fisher BioReagents
BP1341-4 |
4 L | Poly CUBE |
115.00 €
4 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Tris-Glycine-SDS, 10X Solution (Electrophoresis)
Tris-Glycine-SDS is commonly used as a buffer in SDS-PAGE of proteins.Specifications
Tris-Glycine-SDS | |
77-86-1, 7732-18-5, 56-40-6, 151-21-3 | |
Liquid | |
Filtered through a 0.2-micron filter. | |
Pass Test | |
TG-SDS | |
0.25M Tris Base, 1.92M Glycine, and 1.0% (w/v) SDS. |
10X Solution | |
Undesignated | |
DNase free | |
DNase-, RNase- and Protease-Free | |
Protease free | |
Electrophoresis |
Safety and Handling
Recommended Storage : RT
Product Identifier
- TG-SDS 10X Solution
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title