missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tris Hydrochloride (Small White Flakes/Molecular Biology), Fisher BioReagents
For electrophoresis, molecular biology, and cell culture applications
141.00 € - 185.72 €
Chemical Identifiers
CAS | 1185-53-1 |
---|---|
Molecular Formula | C4H12ClNO3 |
Molecular Weight (g/mol) | 157.594 |
InChI Key | QKNYBSVHEMOAJP-UHFFFAOYSA-N |
Synonym | Tris(hydroxymethyl)aminomethane Hydrochloride |
PubChem CID | 93573 |
IUPAC Name | 2-amino-2-(hydroxymethyl)propane-1,3-diol;hydrochloride |
SMILES | C(C(CO)(CO)N)O.Cl |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10316893
|
Fisher BioReagents
BP153-500 |
500 g | Poly Bottle |
185.72 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10001223
|
Fisher BioReagents
BP153-1 |
1 kg | Poly Bottle |
141.00 €
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
For electrophoresis, molecular biology, and cell culture applications
Tris Hydrochloride is a buffering medium for electrophoresis, molecular biology, and cell culture applications. It may be combined with Tris Base to simplify Tris buffer preparation.Chemical Identifiers
1185-53-1 | |
157.594 | |
Tris(hydroxymethyl)aminomethane Hydrochloride | |
2-amino-2-(hydroxymethyl)propane-1,3-diol;hydrochloride |
C4H12ClNO3 | |
QKNYBSVHEMOAJP-UHFFFAOYSA-N | |
93573 | |
C(C(CO)(CO)N)O.Cl |
Specifications
0.05 max. (1M solution) at 280nm | |
150°C | |
3.5 to 5.0 | |
DNase free | |
Pass Test | |
H2NC(CH2OH)3·HCl | |
Tris(hydroxymethyl)aminomethane Hydrochloride | |
C(C(CO)(CO)N)O.Cl | |
157.594 | |
157.6 | |
DNase-, RNase- and Protease-Free |
1185-53-1 | |
White | |
Solid | |
≥99 % | |
C4H12ClNO3 | |
Protease free | |
QKNYBSVHEMOAJP-UHFFFAOYSA-N | |
2-amino-2-(hydroxymethyl)propane-1,3-diol;hydrochloride | |
93573 | |
≥99% | |
Tris Hydrochloride |
Safety and Handling

Recommended Storage : Keep in a dry, cool and well-ventilated place. Keep container tightly closed.
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Tris hydrochloride
Signal Word
- Warning
Hazard Category
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Tris Hydrochloride (Small White Flakes/Molecular Biology), Fisher BioReagents