Learn More
Tris(hydroxymethyl)aminomethane, 99.8%, ACS reagent, Thermo Scientific Chemicals
33.06 € - 342.71 €
Chemical Identifiers
CAS | 77-86-1 |
---|---|
Molecular Formula | C4H11NO3 |
Molecular Weight (g/mol) | 121.136 |
MDL Number | MFCD00004679 |
InChI Key | LENZDBCJOHFCAS-UHFFFAOYSA-N |
Synonym | 2-Amino-2-(hydroxymethyl)-1, 3-propanediol, TRIS, Tris buffer, Tromethamine, Tromethane |
PubChem CID | 6503 |
ChEBI | CHEBI:9754 |
IUPAC Name | 2-amino-2-(hydroxymethyl)propane-1,3-diol |
SMILES | C(C(CO)(CO)N)O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10731072
|
Thermo Scientific Acros
424571000 |
100 g | Plastic bottle |
33.06 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10424521
|
Thermo Scientific Acros
424575000 |
500 g | Plastic bottle |
102.51 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11985281
|
Thermo Scientific Acros
424570025 |
2.5 kg | Plastic bottle |
342.71 €
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Biological bufferChemical Identifiers
77-86-1 | |
121.136 | |
LENZDBCJOHFCAS-UHFFFAOYSA-N | |
6503 | |
2-amino-2-(hydroxymethyl)propane-1,3-diol |
C4H11NO3 | |
MFCD00004679 | |
2-Amino-2-(hydroxymethyl)-1, 3-propanediol, TRIS, Tris buffer, Tromethamine, Tromethane | |
CHEBI:9754 | |
C(C(CO)(CO)N)O |
Specifications
0.2 max. at 290nm, (40% aq. soln.) (1 cm cell), (40% aq. soln.) (1 cm cell) at 0nm | |
77-86-1 | |
219.0°C to 220.0°C (10.0mmHg) | |
Crystalline Powder, Crystals and/or Chunks | |
Authentic | |
(HOCH2)3CNH2 | |
04, 303 | |
2-Amino-2-(hydroxymethyl)-1, 3-propanediol, TRIS, Tris buffer, Tromethamine, Tromethane | |
C(C(CO)(CO)N)O | |
121.136 | |
CHEBI:9754 | |
99.8% | |
Tris(hydroxymethyl)aminomethane |
(40% aq. soln.) (1 cm cell) | |
>168.5°C | |
White | |
99.8% | |
C4H11NO3 | |
MFCD00004679 | |
15, 9951 | |
LENZDBCJOHFCAS-UHFFFAOYSA-N | |
2-amino-2-(hydroxymethyl)propane-1,3-diol | |
6503 | |
121.14 | |
ACS Reagent |
Safety and Handling
GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 201-064-4
missing translation for 'rtecsNumber' : TY2900000
missing translation for 'tsca' : TSCA
- Tris(hydroxymethyl)aminomethane
RUO – Research Use Only