missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Tris(hydroxymethyl)aminomethane acetate salt, 99%

Product Code. 11905291
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Plastic bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Biological buffer
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6850-28-8
Molecular Formula C4H11NO3·C2H4O2
Molecular Weight (g/mol) 181.19
InChI Key PIEPQKCYPFFYMG-UHFFFAOYSA-N
Synonym tris hydroxymethyl aminomethane acetate, tris acetate, tris acetate salt, 2-amino-2-hydroxymethyl propane-1,3-diol acetate salt, trizma acetate, tris hydroxymethyl aminomethane acetate salt, 2-hydroxy-1,1-bis hydroxymethyl ethyl ammonium acetate, 1,3-dihydroxy-2-hydroxymethyl propan-2-aminium acetate, acetic acid; tris buffer, tris-acetate
PubChem CID 16218782
ChEBI CHEBI:66869
IUPAC Name acetic acid;2-amino-2-(hydroxymethyl)propane-1,3-diol
SMILES CC(=O)O.C(C(CO)(CO)N)O

Specifications

Absorbance 0.5 max. (c=50, water) at 290nm
Color White
Assay Percent Range 99%
Packaging Plastic bottle
Quantity 100 g
Solubility Information (1g/ml in water) Clear colorless
Formula Weight 181.19
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material Tris(hydroxymethyl)aminomethane acetate salt

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'