missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Tris(hydroxymethyl)aminomethane acetate salt, 99%
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Plastic bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Biological buffer
Specifications
Specifications
Absorbance | 0.5 max. (c=50, water) at 290nm |
Color | White |
Assay Percent Range | 99% |
Packaging | Plastic bottle |
Quantity | 25 g |
Solubility Information | (1g/ml in water) Clear colorless |
Formula Weight | 181.19 |
Percent Purity | 99% |
Physical Form | Crystalline Powder |
Chemical Name or Material | Tris(hydroxymethyl)aminomethane acetate salt |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'