Learn More
Thermo Scientific Chemicals Tris(hydroxymethyl)aminomethane hydrochloride, 99+%
Description
Tris(hydroxymethyl)aminomethane hydrochloride is used as a component of buffer solutions in biology, biochemistry and molecular biology. It is also used for crystallization of proteins at various pH values. Further, it inhibits motor responses to adrenergic motor nerve stimulation in smooth muscles. In addition to this, it is used in coelomic fluid and in cortland medium to store unfertilized rainbow trout eggs.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Tris(hydroxymethyl)aminomethane hydrochloride is used as a component of buffer solutions in biology, biochemistry and molecular biology. It is also used for crystallization of proteins at various pH values. Further, it inhibits motor responses to adrenergic motor nerve stimulation in smooth muscles. In addition to this, it is used in coelomic fluid and in cortland medium to store unfertilized rainbow trout eggs.
Solubility
Soluble in water.
Notes
Hygroscopic. Incompatible with strong oxidizing agents, strong acids and bases.

Specifications
Specifications
Melting Point | ∼152°C (decomposition) |
pH | 3.5 to 5.0 |
Linear Formula | (HOCH2)3CNH2·HCl |
Quantity | 1000 g |
Beilstein | 3675235 |
Sensitivity | Hygroscopic |
Solubility Information | Soluble in water. |
Formula Weight | 157.6 |
Percent Purity | ≥99% |
Chemical Name or Material | Tris (hydroxymethyl)aminomethane hydrochloride |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.