missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Tris(hydroxymethyl)aminomethane, Molecular Biology Grade, 99.9%

Product Code. 15460167
Click to view available options
Quantity:
100 g
500 g

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 77-86-1
Molecular Formula C4H11NO3
Molecular Weight (g/mol) 121.136
MDL Number MFCD00004679
InChI Key LENZDBCJOHFCAS-UHFFFAOYSA-N
Synonym TRIS
PubChem CID 6503
ChEBI CHEBI:9754
IUPAC Name 2-amino-2-(hydroxymethyl)propane-1,3-diol
SMILES C(C(CO)(CO)N)O

Specifications

Melting Point 168°C to 172°C
Boiling Point 219°C to 220°C (10mmHg)
Color White
Density 1.353
Physical Form Crystalline Powder
Quantity 100 g
Beilstein 741883
Merck Index 14,9772
Solubility Information Freely soluble in water
Formula Weight 121.14
Percent Purity 99.9%
Grade Molecular Biology
Chemical Name or Material Tris(hydroxymethyl)aminomethane
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.