missing translation for 'onlineSavingsMsg'
Learn More

Tris(Hydroxymethyl) Methylamine, 'Tris Buffer', 99+%, Extra Pure, SLR, Fisher Chemical™

Chemical Identifiers

CAS 77-86-1
Molecular Formula C4H11NO3
Molecular Weight (g/mol) 121.136
MDL Number 4679
InChI Key LENZDBCJOHFCAS-UHFFFAOYSA-N
PubChem CID 6503
ChEBI CHEBI:9754
IUPAC Name 2-amino-2-(hydroxymethyl)propane-1,3-diol
SMILES C(C(CO)(CO)N)O
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10172590
View Documents Promotion Details
Fisher Chemical
T/3710/48
100 g N/A
N/A
10528830
View Documents Promotion Details
Fisher Chemical
T/3710/50
250 g N/A
N/A
10785341
View Documents Promotion Details
Fisher Chemical
T/3710/60
1 kg N/A
N/A
Description

Description

Tris(Hydroxymethyl) Methylamine, 'Tris Buffer', 99+%, Extra Pure, SLR

Specifications

Chemical Identifiers

77-86-1
121.136
LENZDBCJOHFCAS-UHFFFAOYSA-N
CHEBI:9754
C(C(CO)(CO)N)O
C4H11NO3
4679
6503
2-amino-2-(hydroxymethyl)propane-1,3-diol

Specifications

77-86-1
219°C
10.4
C4H11NO3
Plastic powder jar
C(C(CO)(CO)N)O
121.136
CHEBI:9754
168.5°C
White
Powder or Solid
4679
LENZDBCJOHFCAS-UHFFFAOYSA-N
2-amino-2-(hydroxymethyl)propane-1,3-diol
6503
121.14g/mol
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Tris(hydroxymethyl)aminomethane

SDS
Documents

Documents

Product Certifications