missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tris, MP Biomedicals™
Click to view available options
Quantity:
250 g
500 g
Description
Tris have been useful as buffers in a wide variety of biological systems. It is used in pH control in vitro and in vivo for body fluids and in buffering systems for electrophoresis applications.
- Used in assays used to characterize the activity and kinetics of the enzymes that catalyze SUMOylation of Small ubiquitin-like proteins (SUMO) and SUMO-dependent protein-protein interactions
- Used as a starting material for polymers, oxazolones (with carboxylic acids) and oxazolidines (with aldehydes)
- Does not precipitate calcium salts and is of value in maintaining solubility of manganese salts
- Can be used for the direct standardization of a strong acid solution; the equivalence point can be determined either potentiometrically or by use of a suitable indicator such as 3-(4-Dimethylamino-1-naphthylazo)-4-methoxybenzenesulfonic acid
Specifications
Specifications
Physical Form | Crystalline Powder |
Quantity | 250 g |
Solubility Information | Soluble in water (550mg/mL), ethylene glycol (79.1mg/mL), methanol (26mg/mL), anhydrous ethanol (14.6mg/mL), 95% ethanol (22.0mg/mL), DMF (14mg/mL), acetone (2mg/mL), ethyl acetate (0.5mg/mL), olive oil (0.4mg/mL), and chloroform (0.05mg/mL). |
Formula Weight | 121.14 |
Percent Purity | >99.9% |
Grade | Ultrapure |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'